Anti-SLC8B1 Rabbit Polyclonal Antibody
77799-871
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen name:SLC24A6
- Antigen symbol:SLC8B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6J4K2
- Antigen synonyms:FLJ22233|Na(+)/K(+)/Ca(2+) exchange protein 6|Na(+)/K(+)/Ca(2+)-exchange protein 6|NCKX6|NCKX6_HUMAN|NCLX|Slc24a6|Sodium/potassium/calcium exchanger|Sodium/potassium/calcium exchanger 6|Solute carrier family 24 (sodium/potassium/calcium exchanger) member 6|Solute carrier family 24 member 6
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: SLC8B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human