Anti-SERPINB1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: SERPINB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77799-129
- 77799-130
- Antigen symbol:
- SERPINB1
- SERPINB1
- Antigen name:
- Leukocyte elastase inhibitor protein (100-379AA)
- Leukocyte elastase inhibitor protein (100-379AA)
- Antigen synonyms:
- Anti elastase | EI| ELANH2 | Epididymis luminal protein 57| HEL57| ILEU_HUMAN| LEI| Leukocyte elastase inhibitor| M/NEI| MNEI | Monocyte/neutrophil elastase inhibitor| Peptidase inhibitor 2| Pi-2| PI2 | Serine (or cysteine) proteinase inhibitor clade B (ovalbumin) member 1| Serpin B1| Serpin peptidase inhibitor clade B (ovalbumin) member 1| serpinb1
- Anti elastase | EI| ELANH2 | Epididymis luminal protein 57| HEL57| ILEU_HUMAN| LEI| Leukocyte elastase inhibitor| M/NEI| MNEI | Monocyte/neutrophil elastase inhibitor| Peptidase inhibitor 2| Pi-2| PI2 | Serine (or cysteine) proteinase inhibitor clade B (ovalbumin) member 1| Serpin B1| Serpin peptidase inhibitor clade B (ovalbumin) member 1| serpinb1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30740
- P30740
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together