Anti-FANCD2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: FANCD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77836-902
- 77836-901
- Antigen symbol:
- FANCD2
- FANCD2
- Antigen name:
- Fanconi anemia group D2 protein (1-291AA)
- Fanconi anemia group D2 protein (1-291AA)
- Antigen synonyms:
- DKFZp762A223| FA 4| FA D2| FA4| FAC D2| FACD 2| FACD| FACD2| FACD2_HUMAN| FAD| FAD2| FANC D2| FANCD 2| FANCD| FANCD2| FANCONI ANEMIA COMPLEMENTATION GROUP D| Fanconi anemia complementation group D2| Fanconi anemia group D2 protein| FANCONI PANCYTOPENIA TYPE 4| FLJ23826| OTTHUMP00000158853| OTTHUMP00000207925| Protein FACD2| Type 4 Fanconi pancytopenia
- DKFZp762A223| FA 4| FA D2| FA4| FAC D2| FACD 2| FACD| FACD2| FACD2_HUMAN| FAD| FAD2| FANC D2| FANCD 2| FANCD| FANCD2| FANCONI ANEMIA COMPLEMENTATION GROUP D| Fanconi anemia complementation group D2| Fanconi anemia group D2 protein| FANCONI PANCYTOPENIA TYPE 4| FLJ23826| OTTHUMP00000158853| OTTHUMP00000207925| Protein FACD2| Type 4 Fanconi pancytopenia
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXW9
- Q9BXW9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together