Anti-SUS1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SUS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharum officinarum
Specifications
- Catalog No:
- 77816-110
- 77816-111
- Antigen symbol:
- SUS1
- SUS1
- Antigen name:
- Recombinant Saccharum officinarum Sucrose synthase protein (1-218AA)
- Recombinant Saccharum officinarum Sucrose synthase protein (1-218AA)
- Antigen synonyms:
- SUS1| Sucrose synthase| EC 2.4.1.13| Sucrose-UDP glucosyltransferase| Fragment
- SUS1| Sucrose synthase| EC 2.4.1.13| Sucrose-UDP glucosyltransferase| Fragment
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Saccharum officinarum
- Saccharum officinarum
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31925
- P31925
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together