Anti-GNB4 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNB4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-692
- 77835-693
- Antigen symbol:
- GNB4
- GNB4
- Antigen name:
- Guanine nucleotide-binding protein subunit beta-4 protein (105-185AA)
- Guanine nucleotide-binding protein subunit beta-4 protein (105-185AA)
- Antigen synonyms:
- G protein beta 4 subunit|GBB4_HUMAN|GNB 4|Guanine nucleotide binding protein (G protein) beta polypeptide 4|Guanine nucleotide binding protein beta 4 subunit|Guanine nucleotide binding protein beta polypeptide 4|Guanine nucleotide binding protein beta subunit 4|Guanine nucleotide binding protein subunit beta 4|Guanine nucleotide-binding protein subunit beta-4|Transducin beta chain 4
- G protein beta 4 subunit|GBB4_HUMAN|GNB 4|Guanine nucleotide binding protein (G protein) beta polypeptide 4|Guanine nucleotide binding protein beta 4 subunit|Guanine nucleotide binding protein beta polypeptide 4|Guanine nucleotide binding protein beta subunit 4|Guanine nucleotide binding protein subunit beta 4|Guanine nucleotide-binding protein subunit beta-4|Transducin beta chain 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HAV0
- Q9HAV0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together