Anti-LYPLA2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: LYPLA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77808-372
- 77808-373
- Antigen symbol:
- LYPLA2
- LYPLA2
- Antigen name:
- Acyl-protein thioesterase 2 protein (1-226AA)
- Acyl-protein thioesterase 2 protein (1-226AA)
- Antigen synonyms:
- Acyl protein thioesterase 2|Acyl protein thioesterase|Acyl-protein thioesterase 2|APT 2|APT-2|APT2|DJ886K2.4|EC 3.1.2.-|LPL I|LPL II|LPL-II|LYPA2_HUMAN|LYPLA 2|Lypla2|Lysophospholipase II|LysoPLA II|OTTHUMP00000002999|OTTHUMP00000044831
- Acyl protein thioesterase 2|Acyl protein thioesterase|Acyl-protein thioesterase 2|APT 2|APT-2|APT2|DJ886K2.4|EC 3.1.2.-|LPL I|LPL II|LPL-II|LYPA2_HUMAN|LYPLA 2|Lypla2|Lysophospholipase II|LysoPLA II|OTTHUMP00000002999|OTTHUMP00000044831
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95372
- O95372
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together