Anti-SWAP70 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: SWAP70
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77842-720
- 77842-719
- Antigen symbol:
- SWAP70
- SWAP70
- Antigen name:
- Switch-associated protein 70 protein (1-300AA)
- Switch-associated protein 70 protein (1-300AA)
- Antigen synonyms:
- KIAA0640|70kDa|AV235546|FLJ39540|HSPC321|SWA 70 protein|SWAP 70|SWAP switching B-cell complex 70kDa subunit|SWAP-70|SWAP70|Switch associated protein 70|Switch-associated protein 70|SWP70_HUMAN
- KIAA0640|70kDa|AV235546|FLJ39540|HSPC321|SWA 70 protein|SWAP 70|SWAP switching B-cell complex 70kDa subunit|SWAP-70|SWAP70|Switch associated protein 70|Switch-associated protein 70|SWP70_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UH65
- Q9UH65
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together