Anti-RTN4RL2 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RTN4RL2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-685
- 77832-684
- Antigen symbol:
- RTN4RL2
- RTN4RL2
- Antigen name:
- Reticulon-4 receptor-like 2 protein (355-372AA)
- Reticulon-4 receptor-like 2 protein (355-372AA)
- Antigen synonyms:
- RTN4RL2|NGRH1|NGRL3|Reticulon-4 receptor-like 2|Nogo receptor-like 3|Nogo-66 receptor homolog 1|Nogo-66 receptor-related protein 2|NgR2
- RTN4RL2|NGRH1|NGRL3|Reticulon-4 receptor-like 2|Nogo receptor-like 3|Nogo-66 receptor homolog 1|Nogo-66 receptor-related protein 2|NgR2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UN3
- Q86UN3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together