Anti-TRIM36 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM36
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-391
- 77837-390
- Antigen symbol:
- TRIM36
- TRIM36
- Antigen name:
- E3 ubiquitin-protein ligase TRIM36 protein (70-280AA)
- E3 ubiquitin-protein ligase TRIM36 protein (70-280AA)
- Antigen synonyms:
- Acrosome RBCC protein|E3 ubiquitin protein ligase TRIM36|E3 ubiquitin-protein ligase Trim36|Haprin|RBCC728|RING finger protein 98|RNF98|TRI36_HUMAN|TRIM 36|Trim36|Tripartite motif containing 36|Tripartite motif containing protein 36|Tripartite motif protein 36|Tripartite motif-containing protein 36|Zinc binding protein Rbcc728|Zinc-binding protein Rbcc728
- Acrosome RBCC protein|E3 ubiquitin protein ligase TRIM36|E3 ubiquitin-protein ligase Trim36|Haprin|RBCC728|RING finger protein 98|RNF98|TRI36_HUMAN|TRIM 36|Trim36|Tripartite motif containing 36|Tripartite motif containing protein 36|Tripartite motif protein 36|Tripartite motif-containing protein 36|Zinc binding protein Rbcc728|Zinc-binding protein Rbcc728
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQ86
- Q9NQ86
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together