Anti-PNCK Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PNCK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-617
- 77830-616
- Antigen symbol:
- PNCK
- PNCK
- Antigen name:
- Calcium/calmodulin-dependent protein kinase type 1B protein (1-115AA)
- Calcium/calmodulin-dependent protein kinase type 1B protein (1-115AA)
- Antigen synonyms:
- Calcium/calmodulin dependent protein kinase type 1B|Calcium/calmodulin-dependent protein kinase type 1B|CaM KI beta|CaM kinase I beta|CaM kinase IB|CaM-KI beta|CaMK1b|CaMKI beta|CaMKI-beta|EC 2.7.11.17|KCC1B_HUMAN|MGC45419|PNCK|Pregnancy up regulated non ubiquitously expressed CaM kinase|Pregnancy up-regulated non-ubiquitously-expressed CaM kinase
- Calcium/calmodulin dependent protein kinase type 1B|Calcium/calmodulin-dependent protein kinase type 1B|CaM KI beta|CaM kinase I beta|CaM kinase IB|CaM-KI beta|CaMK1b|CaMKI beta|CaMKI-beta|EC 2.7.11.17|KCC1B_HUMAN|MGC45419|PNCK|Pregnancy up regulated non ubiquitously expressed CaM kinase|Pregnancy up-regulated non-ubiquitously-expressed CaM kinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P2M8
- Q6P2M8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together