Anti-RBP3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RBP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-848
- 77796-847
- Antigen symbol:
- RBP3
- RBP3
- Antigen name:
- Retinol-binding protein 3 protein (1146-1246AA)
- Retinol-binding protein 3 protein (1146-1246AA)
- Antigen synonyms:
- D10S64|D10S65|D10S66|Interphotoreceptor retinoid binding protein|Interphotoreceptor retinoid-binding protein|Interstitial retinol binding protein|Interstitial retinol-binding protein|IRBP|RBP 3|rbp3|RBPI|RET3_HUMAN|Retinol binding protein 3|Retinol binding protein 3 interstitial|Retinol-binding protein 3|RP66
- D10S64|D10S65|D10S66|Interphotoreceptor retinoid binding protein|Interphotoreceptor retinoid-binding protein|Interstitial retinol binding protein|Interstitial retinol-binding protein|IRBP|RBP 3|rbp3|RBPI|RET3_HUMAN|Retinol binding protein 3|Retinol binding protein 3 interstitial|Retinol-binding protein 3|RP66
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10745
- P10745
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together