Anti-TNFRSF13C Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TNFRSF13C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-221
- 77833-222
- Antigen symbol:
- TNFRSF13C
- TNFRSF13C
- Antigen name:
- Tumor necrosis factor receptor superfamily member 13C protein (1-78AA)
- Tumor necrosis factor receptor superfamily member 13C protein (1-78AA)
- Antigen synonyms:
- B cell activating factor receptor|B-cell-activating factor receptor|BAFF R|BAFF receptor|BAFF-R|BAFFR|BLyS receptor 3|BlySR3|BR 3|BR3|BROMIX|CD 268|CD268|CD268 antigen|CVID4|MGC138235|OTTHUMP00000028746|Prolixin|TNFRSF 13C|Tnfrsf13c|TR13C_HUMAN|Tumor necrosis factor receptor subunit member 13C|Tumor necrosis factor receptor superfamily member 13C
- B cell activating factor receptor|B-cell-activating factor receptor|BAFF R|BAFF receptor|BAFF-R|BAFFR|BLyS receptor 3|BlySR3|BR 3|BR3|BROMIX|CD 268|CD268|CD268 antigen|CVID4|MGC138235|OTTHUMP00000028746|Prolixin|TNFRSF 13C|Tnfrsf13c|TR13C_HUMAN|Tumor necrosis factor receptor subunit member 13C|Tumor necrosis factor receptor superfamily member 13C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96RJ3
- Q96RJ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together