Order Entry
Canada
ContactUsLinkComponent
 
Anti-POLR3B Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
 
undefined
Anti-POLR3B Rabbit Recombinant Polyclonal Antibody (HRP)
New Product

 

  • Catalog No:
  • 77842-461
  • 77842-460
  • Antigen symbol:
  • POLR3B
  • POLR3B
  • Antigen name:
  • DNA-directed RNA polymerase III subunit RPC2 protein (831-939AA)
  • DNA-directed RNA polymerase III subunit RPC2 protein (831-939AA)
  • Antigen synonyms:
  • C128|DNA directed RNA polymerase III 127.6 kDa polypeptide|DNA directed RNA polymerase III subunit B|DNA-directed RNA polymerase III 127.6 kDa polypeptide|DNA-directed RNA polymerase III subunit B|DNA-directed RNA polymerase III subunit RPC2|FLJ10388|polr3b|Polymerase (RNA) III (DNA directed) polypeptide B|RNA polymerase III subunit C2|RNA polymerase III subunit RPC2|RPC2|RPC2_HUMAN
  • C128|DNA directed RNA polymerase III 127.6 kDa polypeptide|DNA directed RNA polymerase III subunit B|DNA-directed RNA polymerase III 127.6 kDa polypeptide|DNA-directed RNA polymerase III subunit B|DNA-directed RNA polymerase III subunit RPC2|FLJ10388|polr3b|Polymerase (RNA) III (DNA directed) polypeptide B|RNA polymerase III subunit C2|RNA polymerase III subunit RPC2|RPC2|RPC2_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9NW08
  • Q9NW08
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together