Anti-BUD13 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: BUD13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-812
- 77835-813
- Antigen symbol:
- BUD13
- BUD13
- Antigen name:
- BUD13 homolog protein (132-233AA)
- BUD13 homolog protein (132-233AA)
- Antigen synonyms:
- BUD13 homolog (S. cerevisiae)|BUD13 homolog|BUD13_HUMAN|Cwc26|FLJ27090|fSAP71|Functional spliceosome-associated protein 71|MGC13125|OTTHUMP00000042639|OTTHUMP00000042640
- BUD13 homolog (S. cerevisiae)|BUD13 homolog|BUD13_HUMAN|Cwc26|FLJ27090|fSAP71|Functional spliceosome-associated protein 71|MGC13125|OTTHUMP00000042639|OTTHUMP00000042640
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BRD0
- Q9BRD0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together