Anti-MED28 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MED28
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-096
- 77836-097
- Antigen symbol:
- MED28
- MED28
- Antigen name:
- Mediator of RNA polymerase II transcription subunit 28 protein (1-178AA)
- Mediator of RNA polymerase II transcription subunit 28 protein (1-178AA)
- Antigen synonyms:
- EG1|Endothelial-derived protein 1|FKSG20|Magicin|med28|MED28_HUMAN|Mediator complex subunit 28|Mediator of RNA polymerase II transcription subunit 28|Merlin and Grb2-interacting cytoskeletal protein|Tumor angiogenesis marker EG-1
- EG1|Endothelial-derived protein 1|FKSG20|Magicin|med28|MED28_HUMAN|Mediator complex subunit 28|Mediator of RNA polymerase II transcription subunit 28|Merlin and Grb2-interacting cytoskeletal protein|Tumor angiogenesis marker EG-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H204
- Q9H204
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together