Anti-P4HA1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: P4HA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-077
- 77793-076
- Antigen symbol:
- P4HA1
- P4HA1
- Antigen name:
- Prolyl 4-hydroxylase subunit alpha-1 protein (256-525AA)
- Prolyl 4-hydroxylase subunit alpha-1 protein (256-525AA)
- Antigen synonyms:
- 2-oxoglutarate-4-dioxygenase subunit alpha-1|4-hydroxylase, alpha I subunit|4-PH alpha-1|P4HA|P4HA1_HUMAN|Procollagen-proline|Procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide I|Prolyl 4-hydroxylase subunit alpha-1
- 2-oxoglutarate-4-dioxygenase subunit alpha-1|4-hydroxylase, alpha I subunit|4-PH alpha-1|P4HA|P4HA1_HUMAN|Procollagen-proline|Procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide I|Prolyl 4-hydroxylase subunit alpha-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13674
- P13674
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together