Anti-RPL28 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RPL28
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-926
- 77797-925
- Antigen symbol:
- RPL28
- RPL28
- Antigen name:
- 60S ribosomal protein L28 protein (2-137AA)
- 60S ribosomal protein L28 protein (2-137AA)
- Antigen synonyms:
- 60S ribosomal protein L28|FLJ 43307|FLJ43307|L 28|L28|Ribosomal protein L28|RL28_HUMAN|RPL 28|rpl28
- 60S ribosomal protein L28|FLJ 43307|FLJ43307|L 28|L28|Ribosomal protein L28|RL28_HUMAN|RPL 28|rpl28
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P46779
- P46779
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together