Anti-TFCP2 Rabbit Recombinant Antibody
Catalog # 77824-237
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Alpha-globin transcription factor CP2 protein (1-280AA)
- Antigen symbol:TFCP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q12800
- Antigen synonyms:Alpha globin transcription factor CP2|Alpha-globin transcription factor CP2|CP2|Late SV40 factor|LBP 1C|LBP1C|LSF|LSF1D|SAA3 enhancer factor|SEF|TFCP2|TFCP2_HUMAN|TFCP2C|Transcription factor CP2|Transcription factor LSF
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: TFCP2
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse