Anti-Snrnp35 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Snrnp35
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77841-871
- 77841-872
- Antigen symbol:
- Snrnp35
- Snrnp35
- Antigen name:
- Recombinant Mouse U11/U12 small nuclear ribonucleoprotein 35 kDa protein (1-244AA)
- Recombinant Mouse U11/U12 small nuclear ribonucleoprotein 35 kDa protein (1-244AA)
- Antigen synonyms:
- Snrnp35|U1snrnpbpU11/U12 small nuclear ribonucleoprotein 35 kDa protein|U11/U12 snRNP 35 kDa protein|U1 snRNP-binding protein homolog
- Snrnp35|U1snrnpbpU11/U12 small nuclear ribonucleoprotein 35 kDa protein|U11/U12 snRNP 35 kDa protein|U1 snRNP-binding protein homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9D384
- Q9D384
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together