Anti-Nqo2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: Nqo2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Mouse
- Catalog No:
- 77828-329
- 77828-330
- Antigen symbol:
- Nqo2
- Nqo2
- Antigen name:
- Recombinant Rat Ribosyldihydronicotinamide dehydrogenase [quinone] protein (1-231AA)
- Recombinant Rat Ribosyldihydronicotinamide dehydrogenase [quinone] protein (1-231AA)
- Antigen synonyms:
- Ribosyldihydronicotinamide dehydrogenase [quinone]|EC 1.10.5.1|NRH dehydrogenase [quinone] 2|NRH:quinone oxidoreductase 2|Quinone reductase 2|QR2
- Ribosyldihydronicotinamide dehydrogenase [quinone]|EC 1.10.5.1|NRH dehydrogenase [quinone] 2|NRH:quinone oxidoreductase 2|Quinone reductase 2|QR2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Rat, Mouse
- Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6AY80
- Q6AY80
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together