Anti-RPS27 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: RPS27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77818-586
- 77818-587
- Antigen symbol:
- RPS27
- RPS27
- Antigen name:
- Fusion protein of RPS27
- Fusion protein of RPS27
- Antigen synonyms:
- 2610206D02Rik|3200001M24Rik|40S ribosomal protein S27|C530005D02Rik|Metallopan stimulin 1|Metallopan-stimulin 1|Metallopanstimulin 1|Metallopanstimulin1|MPS 1|MPS-1|MPS1|pmn|Ribosomal protein S27|Ribosomal protein S27 metallopanstimulin 1|RS27_HUMAN|S27|snoRNA MBI 112
- 2610206D02Rik|3200001M24Rik|40S ribosomal protein S27|C530005D02Rik|Metallopan stimulin 1|Metallopan-stimulin 1|Metallopanstimulin 1|Metallopanstimulin1|MPS 1|MPS-1|MPS1|pmn|Ribosomal protein S27|Ribosomal protein S27 metallopanstimulin 1|RS27_HUMAN|S27|snoRNA MBI 112
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P42677
- P42677
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together