Anti-QSOX1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: QSOX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-302
- 77796-301
- Antigen symbol:
- QSOX1
- QSOX1
- Antigen name:
- Sulfhydryl oxidase 1 protein (101-175AA)
- Sulfhydryl oxidase 1 protein (101-175AA)
- Antigen synonyms:
- FLJ34858|hQSOX|Human quiescin (Q6) mRNA|partial cds|Q6|QSCN6|QSOX1|QSOX1_HUMAN|Quiescin Q6|Quiescin Q6 sulfhydryl oxidase 1|Skin sulfhydryl oxidase|Sox|Sulfhydryl oxidase 1|Sulfhydryl oxidase 1 precursor
- FLJ34858|hQSOX|Human quiescin (Q6) mRNA|partial cds|Q6|QSCN6|QSOX1|QSOX1_HUMAN|Quiescin Q6|Quiescin Q6 sulfhydryl oxidase 1|Skin sulfhydryl oxidase|Sox|Sulfhydryl oxidase 1|Sulfhydryl oxidase 1 precursor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00391
- O00391
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together