Order Entry
Canada
ContactUsLinkComponent
 
Anti-TFB2M Rabbit Recombinant Polyclonal Antibody
New Product
 
undefined
Anti-TFB2M Rabbit Recombinant Polyclonal Antibody
New Product

 

  • Catalog No:
  • 77837-191
  • 77837-190
  • Antigen symbol:
  • N''-adenosyl(rRNA) dimethyltransferase 2;S adenosylmethionine 6 N', N' adenosyl(rRNA) dimethyltransferase 2;Dimethyladenosine transferase 2;dimethyladenosine transferase 2 mitochondrial;FLJ22661;FLJ23182;h mtTFB;h mtTFB2;h-mtTFB;h-mtTFB2;HCV NS5A transactivated protein 5;HCV NS5A-transactivated protein 5;hepatitis C virus NS5A transactivated protein 5;Hepatitis C virus NS5A-transactivated protein 5;Hkp1;hTFB2M;Mitochondrial 12S rRNA dimethylase 2;mitochondrial;Mitochondrial dimethyladenosine transferase 2;Mitochondrial transcription factor B2;mtTFB;mtTFB2;NS5ATP5;S-adenosylmethionine-6-N'';TFB2M;TFB2M_HUMAN;Transcription Factor B2;Transcription factor B2 mitochondrial
  • N''-adenosyl(rRNA) dimethyltransferase 2;S adenosylmethionine 6 N', N' adenosyl(rRNA) dimethyltransferase 2;Dimethyladenosine transferase 2;dimethyladenosine transferase 2 mitochondrial;FLJ22661;FLJ23182;h mtTFB;h mtTFB2;h-mtTFB;h-mtTFB2;HCV NS5A transactivated protein 5;HCV NS5A-transactivated protein 5;hepatitis C virus NS5A transactivated protein 5;Hepatitis C virus NS5A-transactivated protein 5;Hkp1;hTFB2M;Mitochondrial 12S rRNA dimethylase 2;mitochondrial;Mitochondrial dimethyladenosine transferase 2;Mitochondrial transcription factor B2;mtTFB;mtTFB2;NS5ATP5;S-adenosylmethionine-6-N'';TFB2M;TFB2M_HUMAN;Transcription Factor B2;Transcription factor B2 mitochondrial
  • Antigen name:
  • Dimethyladenosine transferase 2, mitochondrial protein (30-108AA)
  • Dimethyladenosine transferase 2, mitochondrial protein (30-108AA)
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9H5Q4
  • Q9H5Q4
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together