Anti-ENO1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ENO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-213
- 77803-213
- Antigen symbol:
- ENO1
- ENO1
- Antigen name:
- Alpha-enolase protein (2-428AA)
- Alpha-enolase protein (2-428AA)
- Antigen synonyms:
- 2 phospho D glycerate hydro lyase|2-phospho-D-glycerate hydro-lyase|Alpha enolase|Alpha enolase like 1|Alpha-enolase|C myc promoter binding protein|C-myc promoter-binding protein|EC 4.2.1.11|ENO1L1|ENOA_HUMAN|Enolase 1 (alpha)|Enolase 1 (alpha) like 1|Enolase 1|Enolase alpha|MBP 1|MBP-1|MBP1|MBPB1|MPB 1|MPB-1|MPB1|MYC promoter binding protein 1|NNE|Non neural enolase|Non-neural enolase|Phosphopyruvate hydratase|Plasminogen binding protein|Plasminogen-binding protein|PPH|Tau crystallin
- 2 phospho D glycerate hydro lyase|2-phospho-D-glycerate hydro-lyase|Alpha enolase|Alpha enolase like 1|Alpha-enolase|C myc promoter binding protein|C-myc promoter-binding protein|EC 4.2.1.11|ENO1L1|ENOA_HUMAN|Enolase 1 (alpha)|Enolase 1 (alpha) like 1|Enolase 1|Enolase alpha|MBP 1|MBP-1|MBP1|MBPB1|MPB 1|MPB-1|MPB1|MYC promoter binding protein 1|NNE|Non neural enolase|Non-neural enolase|Phosphopyruvate hydratase|Plasminogen binding protein|Plasminogen-binding protein|PPH|Tau crystallin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06733
- P06733
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together