Anti-DBNDD2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DBNDD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-805
- 77835-804
- Antigen symbol:
- DBNDD2
- DBNDD2
- Antigen name:
- Dysbindin domain-containing protein 2 protein (1-161AA)
- Dysbindin domain-containing protein 2 protein (1-161AA)
- Antigen synonyms:
- DBNDD2|C20orf35Dysbindin domain-containing protein 2|Casein kinase-1 binding protein|CK1BP|HSMNP1
- DBNDD2|C20orf35Dysbindin domain-containing protein 2|Casein kinase-1 binding protein|CK1BP|HSMNP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BQY9
- Q9BQY9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together