Anti-ABCG8 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ABCG8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-381
- 77839-380
- Antigen symbol:
- ABCG8
- ABCG8
- Antigen name:
- ATP-binding cassette sub-family G member 8 protein (176-413AA)
- ATP-binding cassette sub-family G member 8 protein (176-413AA)
- Antigen synonyms:
- ABCG8|ABCG8_HUMAN|ATP binding cassette sub family G (WHITE) member 8 (sterolin 2) |ATP binding cassette sub family G member 8|ATP binding cassette|sub family G (WHITE)|member 8|ATP-binding cassette sub-family G member 8|GBD4|MGC142217 |Sterolin 2|Sterolin-2|STSL
- ABCG8|ABCG8_HUMAN|ATP binding cassette sub family G (WHITE) member 8 (sterolin 2) |ATP binding cassette sub family G member 8|ATP binding cassette|sub family G (WHITE)|member 8|ATP-binding cassette sub-family G member 8|GBD4|MGC142217 |Sterolin 2|Sterolin-2|STSL
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H221
- Q9H221
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together