Anti-MYBPC1 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:200 to 1:500
Type:
Antigen: MYBPC1
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-051
- 77828-052
- Antigen symbol:
- MYBPC1
- MYBPC1
- Antigen name:
- Myosin-binding protein C, slow-type protein (618-779AA)
- Myosin-binding protein C, slow-type protein (618-779AA)
- Antigen synonyms:
- C protein, skeletal muscle slow isoform|C-protein|MYBPC1|MYBPCC|MYBPCS|Myosin binding protein C, slow type|Myosin-binding protein C|MYPC1_HUMAN|skeletal muscle C protein|skeletal muscle slow isoform|Slow MyBP C|Slow MyBP-C|slow-type
- C protein, skeletal muscle slow isoform|C-protein|MYBPC1|MYBPCC|MYBPCS|Myosin binding protein C, slow type|Myosin-binding protein C|MYPC1_HUMAN|skeletal muscle C protein|skeletal muscle slow isoform|Slow MyBP C|Slow MyBP-C|slow-type
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00872
- Q00872
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together