Anti-SLC1A2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC1A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-807
- 77799-808
- Antigen symbol:
- SLC1A2
- SLC1A2
- Antigen name:
- Excitatory amino acid transporter 2 protein (460-574AA)
- Excitatory amino acid transporter 2 protein (460-574AA)
- Antigen synonyms:
- EAA2_HUMAN|EAAT2|Excitatory amino acid transporter 2|Excitotoxic amino acid transporter 2|Glial high affinity glutamate transporter|GLT 1|GLT1|Glutamate aspartate transporter II|Glutamate transporter 1|Glutamate/aspartate transporter II|Sodium dependent glutamate aspartate transporter 2|Sodium-dependent glutamate/aspartate transporter 2|solute carrier family 1 (glial high affinity glutamate transporter)|member 2|Solute carrier family 1 glial high affinity glutamate transporter member 2|Solute carrier family 1 member 2
- EAA2_HUMAN|EAAT2|Excitatory amino acid transporter 2|Excitotoxic amino acid transporter 2|Glial high affinity glutamate transporter|GLT 1|GLT1|Glutamate aspartate transporter II|Glutamate transporter 1|Glutamate/aspartate transporter II|Sodium dependent glutamate aspartate transporter 2|Sodium-dependent glutamate/aspartate transporter 2|solute carrier family 1 (glial high affinity glutamate transporter)|member 2|Solute carrier family 1 glial high affinity glutamate transporter member 2|Solute carrier family 1 member 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43004
- P43004
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together