Anti-PCMT1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PCMT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-421
- 77793-420
- Antigen symbol:
- PCMT1
- PCMT1
- Antigen name:
- Protein-L-isoaspartate(D-aspartate) O-methyltransferase protein (117-228AA)
- Protein-L-isoaspartate(D-aspartate) O-methyltransferase protein (117-228AA)
- Antigen synonyms:
- PCMT1| Protein-L-isoaspartate(D-aspartate) O-methyltransferase| PIMT| EC 2.1.1.77| L-isoaspartyl protein carboxyl methyltransferase| Protein L-isoaspartyl/D-aspartyl methyltransferase| Protein-beta-aspartate methyltransferase
- PCMT1| Protein-L-isoaspartate(D-aspartate) O-methyltransferase| PIMT| EC 2.1.1.77| L-isoaspartyl protein carboxyl methyltransferase| Protein L-isoaspartyl/D-aspartyl methyltransferase| Protein-beta-aspartate methyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P22061
- P22061
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together