Anti-IMMT Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IMMT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-109
- 77825-108
- Antigen symbol:
- Cell proliferation inducing protein 52;Cell proliferation-inducing gene 4/52 protein;Heart muscle protein;HMP;IMMT;IMMT_HUMAN;Inner membrane protein mitochondrial;Inner mitochrondial membrane;MICOS complex subunit MIC60;MINOS2;Mitochondrial inner membrane organizing system 2;Mitochondrial inner membrane protein;Mitofilin;Motor protein;p87;p87/89;p89;pig4;PIG52;Proliferation-inducing gene 4
- Cell proliferation inducing protein 52;Cell proliferation-inducing gene 4/52 protein;Heart muscle protein;HMP;IMMT;IMMT_HUMAN;Inner membrane protein mitochondrial;Inner mitochrondial membrane;MICOS complex subunit MIC60;MINOS2;Mitochondrial inner membrane organizing system 2;Mitochondrial inner membrane protein;Mitofilin;Motor protein;p87;p87/89;p89;pig4;PIG52;Proliferation-inducing gene 4
- Antigen name:
- MICOS complex subunit MIC60 protein (76-215AA)
- MICOS complex subunit MIC60 protein (76-215AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16891
- Q16891
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together