Anti-AKAP17A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: AKAP17A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77807-640
- 77807-641
- Antigen symbol:
- AKAP17A
- AKAP17A
- Antigen name:
- A-kinase anchor protein 17A protein (1-350AA)
- A-kinase anchor protein 17A protein (1-350AA)
- Antigen synonyms:
- AKAP17A|CXYorf3|DXYS155E|SFRS17A|XE7A-kinase anchor protein 17A|AKAP-17A|721P|B-lymphocyte antigen|Protein XE7|Protein kinase A-anchoring protein 17A|PRKA17A|Splicing factor|arginine/serine-rich 17A
- AKAP17A|CXYorf3|DXYS155E|SFRS17A|XE7A-kinase anchor protein 17A|AKAP-17A|721P|B-lymphocyte antigen|Protein XE7|Protein kinase A-anchoring protein 17A|PRKA17A|Splicing factor|arginine/serine-rich 17A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02040
- Q02040
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together