Anti-UBXN2B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBXN2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-575
- 77822-574
- Antigen symbol:
- UBXN2B
- UBXN2B
- Antigen name:
- UBX domain-containing protein 2B protein (2-250AA)
- UBX domain-containing protein 2B protein (2-250AA)
- Antigen synonyms:
- NSFL1 cofactor p37|p37|p97 cofactor p37|UBX domain containing protein 2B|UBX domain protein 2B|UBX domain-containing protein 2B|UBX2B_HUMAN|UBXN2B
- NSFL1 cofactor p37|p37|p97 cofactor p37|UBX domain containing protein 2B|UBX domain protein 2B|UBX domain-containing protein 2B|UBX2B_HUMAN|UBXN2B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14CS0
- Q14CS0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together