Anti-ZMYND11 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZMYND11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-483
- 77824-482
- Antigen symbol:
- ZMYND11
- ZMYND11
- Antigen name:
- Zinc finger MYND domain-containing protein 11 protein (363-513AA)
- Zinc finger MYND domain-containing protein 11 protein (363-513AA)
- Antigen synonyms:
- ZMYND11|BRAM1|BS69|Zinc finger MYND domain-containing protein 11|Adenovirus 5 E1A-binding protein|Bone morphogenetic protein receptor-associated molecule 1|Protein BS69
- ZMYND11|BRAM1|BS69|Zinc finger MYND domain-containing protein 11|Adenovirus 5 E1A-binding protein|Bone morphogenetic protein receptor-associated molecule 1|Protein BS69
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15326
- Q15326
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together