Anti-FASTKD3 Rabbit Polyclonal Antibody
77818-533
New Product
- Antigen name:Synthesized peptide derived from internal of FAKD3
- Antigen symbol:FASTKD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14CZ7
- Antigen synonyms:FASTKD3FAST kinase domain-containing protein 3|mitochondrial
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: FASTKD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human