Anti-SMARCA4 Rabbit Polyclonal Antibody
Catalog # 77808-135
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the C-terminal region of Brg-1.
- Antigen symbol:SMARCA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P51532
- Antigen synonyms:ATP dependent helicase SMARCA4|ATP-dependent helicase SMARCA4|BAF 190|BAF190|BAF190A|Brahma protein like 1|BRG1|BRG1 associated factor 190A|BRG1 protein|BRG1-associated factor 190A|BRM/SWI2 related gene 1|Global transcription activator homologous sequence|global transcription activator snf2l4|Homeotic gene regulator|hSNF2b|Mitotic growth and transcription activator|MRD16|Nuclear protein GRB1|Protein brahma homolog 1|Protein BRG-1|Protein BRG1|RTPS2|SMARC A4|SMARCA4|SMCA4_HUMAN|SNF2|SNF2 beta|SNF2 like 4|SNF2-beta|SNF2B|SNF2L4|SNF2LB|Sucrose nonfermenting like 4|SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 4|SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4|SWI2|Transcription activator BRG1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: SMARCA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat