Anti-MTFP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MTFP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-749
- 77836-750
- Antigen symbol:
- MTFP1
- MTFP1
- Antigen name:
- Mitochondrial fission process protein 1 protein (1-166AA)
- Mitochondrial fission process protein 1 protein (1-166AA)
- Antigen synonyms:
- HSPC 242|HSPC242|Mitochondrial 18 kDa protein|Mitochondrial fission process protein 1|Mitochondrial fission protein MTP18|Mitochondrial protein 18 kDa|MTFP1|MTFP1_HUMAN|MTP 18|MTP18
- HSPC 242|HSPC242|Mitochondrial 18 kDa protein|Mitochondrial fission process protein 1|Mitochondrial fission protein MTP18|Mitochondrial protein 18 kDa|MTFP1|MTFP1_HUMAN|MTP 18|MTP18
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UDX5
- Q9UDX5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together