Anti-SWI5 Rabbit Recombinant Polyclonal Antibody (Biotin)
New Product
Type:
Antigen: SWI5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77825-652
- 77825-653
- Antigen symbol:
- SWI5;C9orf119;SAE3;DNA repair protein SWI5 homolog;HBV DNAPTP1-transactivated protein A;Protein SAE3 homolog
- SWI5;C9orf119;SAE3;DNA repair protein SWI5 homolog;HBV DNAPTP1-transactivated protein A;Protein SAE3 homolog
- Antigen name:
- DNA repair protein SWI5 homolog protein (54-112AA)
- DNA repair protein SWI5 homolog protein (54-112AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q1ZZU3
- Q1ZZU3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together