Anti-RBBP7 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: RBBP7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-594
- 77824-593
- Antigen symbol:
- G1/S transition control protein binding protein RbAp46;Histone acetyltransferase type B subunit 2;Histone binding protein RBBP7;Histone-binding protein RBBP7;MGC138867;MGC138868;Nucleosome remodeling factor subunit RBAP46;Nucleosome-remodeling factor subunit RBAP46;RBAP46;RBBP 7;RBBP-7;RBBP7;RBBP7_HUMAN;Retinoblastoma binding protein 7;Retinoblastoma binding protein p46;Retinoblastoma-binding protein 7;Retinoblastoma-binding protein p46;Retinoblastoma-binding protein RbAp46
- G1/S transition control protein binding protein RbAp46;Histone acetyltransferase type B subunit 2;Histone binding protein RBBP7;Histone-binding protein RBBP7;MGC138867;MGC138868;Nucleosome remodeling factor subunit RBAP46;Nucleosome-remodeling factor subunit RBAP46;RBAP46;RBBP 7;RBBP-7;RBBP7;RBBP7_HUMAN;Retinoblastoma binding protein 7;Retinoblastoma binding protein p46;Retinoblastoma-binding protein 7;Retinoblastoma-binding protein p46;Retinoblastoma-binding protein RbAp46
- Antigen name:
- Histone-binding protein RBBP7 protein (1-425AA)
- Histone-binding protein RBBP7 protein (1-425AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16576
- Q16576
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together