Anti-ANKRD2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ANKRD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-319
- 77839-320
- Antigen symbol:
- ANKRD2
- ANKRD2
- Antigen name:
- Ankyrin repeat domain-containing protein 2 protein (1-210AA)
- Ankyrin repeat domain-containing protein 2 protein (1-210AA)
- Antigen synonyms:
- ANKRD2|ARPPAnkyrin repeat domain-containing protein 2|Skeletal muscle ankyrin repeat protein|hArpp
- ANKRD2|ARPPAnkyrin repeat domain-containing protein 2|Skeletal muscle ankyrin repeat protein|hArpp
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZV1
- Q9GZV1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together