Anti-UBE2D1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77803-833
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Ubiquitin-conjugating enzyme E2 D1 protein (1-147AA)
- Antigen symbol:UBE2D1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P51668
- Antigen synonyms:E2(17)KB1|SFT|Stimulator of Fe transport|UB2D1_HUMAN|UBC 4/5|UBC4/5|UBC4/5 homolog|UBC4/5, S. cerevisiae, homolog of|UBCH 5|UBCH 5A|UbcH5|UBCH5A|Ube2d1|Ubiquitin carrier protein|Ubiquitin carrier protein D1|Ubiquitin conjugating enzyme E2 17 kDa 1|Ubiquitin conjugating enzyme E2 D1|Ubiquitin conjugating enzyme E2D 1 (UBC4/5 homolog yeast)|Ubiquitin conjugating enzyme E2D 1|Ubiquitin conjugating enzyme UBCH5A|Ubiquitin protein ligase D1|Ubiquitin-conjugating enzyme E2 D1|Ubiquitin-conjugating enzyme E2(17)KB 1|Ubiquitin-conjugating enzyme E2-17 kDa 1|Ubiquitin-protein ligase D1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: UBE2D1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse