Anti-ELOC Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: ELOC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77802-076
- 77802-075
- Antigen symbol:
- ELOC
- ELOC
- Antigen name:
- Elongin-C protein (1-112AA)
- Elongin-C protein (1-112AA)
- Antigen synonyms:
- Elo C|EloC|ELOC_HUMAN|Elongin 15 kDa subunit|Elongin C|Elongin-C|ElonginC|RNA polymerase II transcription factor SIII subunit C|SIII|SIII p15|TCEB 1|tceb1|Transcription elongation factor B (SIII) polypeptide 1|Transcription elongation factor B polypeptide 1
- Elo C|EloC|ELOC_HUMAN|Elongin 15 kDa subunit|Elongin C|Elongin-C|ElonginC|RNA polymerase II transcription factor SIII subunit C|SIII|SIII p15|TCEB 1|tceb1|Transcription elongation factor B (SIII) polypeptide 1|Transcription elongation factor B polypeptide 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15369
- Q15369
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together