Anti-TGOLN2 Rabbit Polyclonal Antibody
77802-447
New Product
- Antigen name:TGOLN2,TGN46
- Antigen symbol:TGOLN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O43493
- Antigen synonyms:TGN 46|TGN 51|TGN38|TGN38 homolog|TGN38B|TGN46|TGN48|TGN51|TGOLN2|TGON2_HUMAN|Trans Golgi network integral membrane protein 2|Trans golgi network protein 2|Trans Golgi network protein TGN51|Trans-Golgi network integral membrane protein 2|Trans-Golgi network protein TGN51|TTGN2
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: TGOLN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human