Anti-FSCN1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FSCN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-787
- 77823-786
- Antigen symbol:
- FSCN1
- FSCN1
- Antigen name:
- Fascin protein (2-187AA)
- Fascin protein (2-187AA)
- Antigen synonyms:
- 55 kDa actin bundling protein|55 kDa actin-bundling protein|Actin bundling protein|actin bundling protein|55-KD|FAN 1|FAN1|Fascin 1|Fascin actin bundling protein 1|Fascin|Fascin homolog 1 actin bundling protein (Strongylocentrotus purpuratus)|Fascin homolog 1|sea urchin|homolog of|1|Fascin1|FLJ38511|FSCN 1|FSCN1|FSCN1_HUMAN|HSN|p55|Singed (Drosophila) like (sea urchin fascin homolog like)|Singed drosophila homolog like|Singed like (fascin homolog sea urchin) (Drosophila)|Singed like (fascin homolog sea urchin)|Singed like protein|Singed|drosophila|Singed-like protein|SNL|Strongylocentrotus purpuratus
- 55 kDa actin bundling protein|55 kDa actin-bundling protein|Actin bundling protein|actin bundling protein|55-KD|FAN 1|FAN1|Fascin 1|Fascin actin bundling protein 1|Fascin|Fascin homolog 1 actin bundling protein (Strongylocentrotus purpuratus)|Fascin homolog 1|sea urchin|homolog of|1|Fascin1|FLJ38511|FSCN 1|FSCN1|FSCN1_HUMAN|HSN|p55|Singed (Drosophila) like (sea urchin fascin homolog like)|Singed drosophila homolog like|Singed like (fascin homolog sea urchin) (Drosophila)|Singed like (fascin homolog sea urchin)|Singed like protein|Singed|drosophila|Singed-like protein|SNL|Strongylocentrotus purpuratus
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16658
- Q16658
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together