Anti-NACA Rabbit Polyclonal Antibody
77797-630
New Product
- Antigen name:Synthesized peptide derived from NACA around the phosphorylation site of S43.
- Antigen symbol:NACA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q13765
- Antigen synonyms:Alpha NAC|Alpha-NAC|Hom s 2.02|HSD48|MGC117224|NAC-alpha|NACA/1.9.2|NACA_HUMAN|NACA1|Nascent polypeptide associated complex alpha polypeptide|Nascent polypeptide associated complex alpha subunit|Nascent polypeptide associated complex subunit alpha|Nascent polypeptide-associated complex subunit alpha
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: NACA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat