Anti-ENOSF1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: ENOSF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-209
- 77830-208
- Antigen symbol:
- ENOSF1
- ENOSF1
- Antigen name:
- Mitochondrial enolase superfamily member 1 protein (171-286AA)
- Mitochondrial enolase superfamily member 1 protein (171-286AA)
- Antigen synonyms:
- Antisense RNA to thymidylate synthase| ENOF1_HUMAN| Enolase superfamily member 1| ENOSF 1| enosf1| HSRTSBETA| Mitochondrial enolase superfamily member 1| RTS alpha| RTS| RTS beta| RTS beta protein| TYMSAS
- Antisense RNA to thymidylate synthase| ENOF1_HUMAN| Enolase superfamily member 1| ENOSF 1| enosf1| HSRTSBETA| Mitochondrial enolase superfamily member 1| RTS alpha| RTS| RTS beta| RTS beta protein| TYMSAS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L5Y1
- Q7L5Y1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together