Anti-USP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: USP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-125
- 77804-126
- Antigen symbol:
- USP2
- USP2
- Antigen name:
- Ubiquitin carboxyl-terminal hydrolase 2 protein (9-265AA)
- Ubiquitin carboxyl-terminal hydrolase 2 protein (9-265AA)
- Antigen synonyms:
- 41 kDa ubiquitin specific protease|41 kDa ubiquitin-specific protease|Deubiquitinating enzyme 2|Ubiquitin carboxyl terminal hydrolase 2|Ubiquitin carboxyl-terminal hydrolase 2|ubiquitin specific peptidase 2|Ubiquitin specific processing protease 2|Ubiquitin specific protease 12|Ubiquitin specific protease 9|Ubiquitin specific protease, 41kD|Ubiquitin thioesterase 2|Ubiquitin thiolesterase 2|Ubiquitin-specific-processing protease 2|Ubiquitin-specific-processing protease testis|UBP-t|UBP2_HUMAN|UBP41|USP9
- 41 kDa ubiquitin specific protease|41 kDa ubiquitin-specific protease|Deubiquitinating enzyme 2|Ubiquitin carboxyl terminal hydrolase 2|Ubiquitin carboxyl-terminal hydrolase 2|ubiquitin specific peptidase 2|Ubiquitin specific processing protease 2|Ubiquitin specific protease 12|Ubiquitin specific protease 9|Ubiquitin specific protease, 41kD|Ubiquitin thioesterase 2|Ubiquitin thiolesterase 2|Ubiquitin-specific-processing protease 2|Ubiquitin-specific-processing protease testis|UBP-t|UBP2_HUMAN|UBP41|USP9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75604
- O75604
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together