Anti-GPR84 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPR84
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-437
- 77836-436
- Antigen symbol:
- GPR84
- GPR84
- Antigen name:
- G-protein coupled receptor 84 protein (202-320AA)
- G-protein coupled receptor 84 protein (202-320AA)
- Antigen synonyms:
- EX 33|EX33|G protein coupled receptor 84|G protein coupled receptor EX 33|G protein coupled receptor EX33|G-protein coupled receptor 84|GPCR 4|GPCR EX33|GPCR4|GPR 84|GPR84|GPR84_HUMAN|Inflammation related G protein coupled receptor EX 33|Inflammation related G protein coupled receptor EX33|Inflammation-related G-protein coupled receptor EX33
- EX 33|EX33|G protein coupled receptor 84|G protein coupled receptor EX 33|G protein coupled receptor EX33|G-protein coupled receptor 84|GPCR 4|GPCR EX33|GPCR4|GPR 84|GPR84|GPR84_HUMAN|Inflammation related G protein coupled receptor EX 33|Inflammation related G protein coupled receptor EX33|Inflammation-related G-protein coupled receptor EX33
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQS5
- Q9NQS5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together