Anti-KRT37/KRT38 Rabbit Polyclonal Antibody
Catalog # 77810-377
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the Internal region of Keratin 37/38
- Antigen symbol:KRT37/KRT38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O76014/O76015
- Antigen synonyms:KRT37|HHA7|HKA7|KRTHA7|Keratin|type I cuticular Ha7|Hair keratin, type I Ha7|Keratin-37|K37|KRT38|HHA8|HKA8|KRTHA8|Keratin, type I cuticular Ha8|Hair keratin, type I Ha8|Keratin-38|K38
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:5000
Type:
Antigen: KRT37/KRT38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human