Order Entry
Canada
ContactUsLinkComponent
Anti-TAS2R14 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-TAS2R14 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-TAS2R14 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77837-621
  • 77837-620
  • Antigen symbol:
  • TAS2R14
  • TAS2R14
  • Antigen name:
  • Taste receptor type 2 member 14 protein (283-317AA)
  • Taste receptor type 2 member 14 protein (283-317AA)
  • Antigen synonyms:
  • T2R14| T2R14_HUMAN| TAS2R14| Taste receptor family B member 1| Taste receptor type 2 member 14| Taste receptor| family B| member 1| type 2| member 14| TRB1
  • T2R14| T2R14_HUMAN| TAS2R14| Taste receptor family B member 1| Taste receptor type 2 member 14| Taste receptor| family B| member 1| type 2| member 14| TRB1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9NYV8
  • Q9NYV8
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together