Anti-mpt63 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: mpt63
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium tuberculosis
- Catalog No:
- 77817-200
- 77817-201
- Antigen symbol:
- mpt63
- mpt63
- Antigen name:
- Recombinant Mycobacterium tuberculosis Immunogenic protein MPT63 protein (30-159AA)
- Recombinant Mycobacterium tuberculosis Immunogenic protein MPT63 protein (30-159AA)
- Antigen synonyms:
- mpt63|Rv1926c|MTCY09F9.38|Immunogenic protein MPT63|16 kDa immunoprotective extracellular protein|Antigen MPT63
- mpt63|Rv1926c|MTCY09F9.38|Immunogenic protein MPT63|16 kDa immunoprotective extracellular protein|Antigen MPT63
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mycobacterium tuberculosis
- Mycobacterium tuberculosis
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P9WIP1
- P9WIP1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together